Diethylenglycolmonohexylether

Lieferant: EHRENSTORFER

Synonyme: 2-(2-(Hexyloxy)ethoxy)ethanol, Diethylene glycol mono-n-hexyl ether

C12606300
EHERC12606300EA 41 EUR
EHERC12606300
Diethylenglycolmonohexylether
Diethylenglycolmonohexylether
Formel: C₁₀H₂₂O₃
Molecular Weight: 190,28 g/mol
MDL: MFCD00010703
CAS-Nummer: 112-59-4
EINECS: 203-988-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR