Digitoxin
Lieferant: EHRENSTORFER
Synonyme:
3β,14β-Dihydroxy-5β-carden-20(22)-olid-3-tridigitoxid
C12633800
EHERC12633800EA
49
EUR
EHERC12633800
Digitoxin
Digitoxin
Formel:
C₄₁H₆₄O₁₃ Molecular Weight: 764.95 g/mol Schmelzpunkt: 276 |
MDL:
MFCD00003686 CAS-Nummer: 71-63-6 EINECS: 200-760-5 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...