o-Dianisidin
Lieferant: EHRENSTORFER
Synonyme:
3,3'-Dimethoxybenzidin
C12721000
EHERC12721000EA
73
EUR
EHERC12721000
o-Dianisidin
o-Dianisidin
Formel:
C₁₄H₁₆N₂O₂ Molecular Weight: 244,29 g/mol |
MDL:
MFCD00008372 CAS-Nummer: 119-90-4 EINECS: 204-355-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...