Dimetridazol
Lieferant: EHRENSTORFER
Synonyme:
1,2-Dimethyl-5-nitroimidazole
C12772000
EHERC12772000EA
46
EUR
EHERC12772000
Dimetridazol
Dimetridazol
Formel:
C₅H₇N₃O₂ Molecular Weight: 141,13 g/mol |
MDL:
MFCD00047046 CAS-Nummer: 551-92-8 EINECS: 209-001-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...