Dimetridazol-[1-Methyl-D3]

Lieferant: EHRENSTORFER

C12772010
EHERC12772010EA 255 EUR
EHERC12772010
Dimetridazol-[1-Methyl-D3]
Dimetridazol-[1-Methyl-D3]
Formel: C₅H₄D₃N₃O₂
MDL: MFCD09265282
CAS-Nummer: 64678-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR