3,5-Dinitro-o-toluamid

Lieferant: EHRENSTORFER

Synonyme: 3,5-Dinitro-2-methylbenzoesäureamid, 3,5-Dinitro-o-tolylsäureamid, Dinitolmid

C12786050
EHERC12786050EA 82 EUR
EHERC12786050
3,5-Dinitro-o-toluamid
3,5-Dinitro-o-toluamid
Formel: C₈H₇N₃O₅
Molecular Weight: 225,16 g/mol
Schmelzpunkt: 180 °C
MDL: MFCD00072065
CAS-Nummer: 148-01-6
EINECS: 205-706-4
Merck Index: 12,03321

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR