Dinobuton (2-sec-Butyl-4,6-dinitrophenylisopropylcarbonat)
Lieferant: EHRENSTORFER
C12790000
EHERC12790000EA
64
EUR
EHERC12790000
Dinobuton (2-sec-Butyl-4,6-dinitrophenylisopropylcarbonat)
Dinobuton (2-sec-Butyl-4,6-dinitrophenylisopropylcarbonat)
Formel:
C₁₄H₁₈N₂O₇ Molecular Weight: 326,31 g/mol |
CAS-Nummer:
973-21-7 EINECS: 213-546-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...