Dinoterb (2-tert-Butyl-4,6-dinitrophenol)

Lieferant: EHRENSTORFER

C12830000
EHERC12830000EA 62 EUR
EHERC12830000
Dinoterb (2-tert-Butyl-4,6-dinitrophenol)
Dinoterb (2-tert-Butyl-4,6-dinitrophenol)
Formel: C₁₀H₁₂N₂O₅
Molecular Weight: 240,22 g/mol
CAS-Nummer: 1420-07-1
EINECS: 215-813-8
UN: 2779
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR