1,4,5,8-Tetraamino-9,10-anthrachinon

Lieferant: EHRENSTORFER

Synonyme: Disperse Blue 1

C12972010
EHERC12972010EA 137 EUR
EHERC12972010
1,4,5,8-Tetraamino-9,10-anthrachinon
1,4,5,8-Tetraamino-9,10-anthrachinon
Formel: C₁₄H₁₂N₄O₂
Molecular Weight: 268,27 g/mol
MDL: MFCD00001225
CAS-Nummer: 2475-45-8
EINECS: 219-603-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR