Disulfiram
Lieferant: EHRENSTORFER
|
Achtung
|
Synonyme:
Tetraethylthiuramdisulfid, Bis(diethyl-thiocarbamoyl)disulfid
C12975000
EHERC12975000EA
45
EUR
EHERC12975000
Disulfiram
Disulfiram
Formel:
C₁₀H₂₀N₂S₄ Molecular Weight: 296,55 g/mol Siedepunkt: 369 °C (1013 hPa) Schmelzpunkt: 70 °C Dichte: 1,34 g/cm³ (20 °C) |
MDL:
MFCD00009048 CAS-Nummer: 97-77-8 EINECS: 202-607-8 UN: 3077 ADR: 9,III Merck Index: 13,03399 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...