Diuron (N-(3,4-Dichlorphenyl)-N',N'-dimethylharnstoff)
Lieferant: EHRENSTORFER
C13020000
EHERC13020000EA
60
EUR
EHERC13020000
Diuron (N-(3,4-Dichlorphenyl)-N',N'-dimethylharnstoff)
Diuron (N-(3,4-Dichlorphenyl)-N',N'-dimethylharnstoff)
Formel:
C₉H₁₀Cl₂N₂O Molecular Weight: 233.10 g/mol |
MDL:
MFCD00018136 CAS-Nummer: 330-54-1 EINECS: 206-354-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...