Erythrosin B Dinatriumsalz

Lieferant: EHRENSTORFER
Achtung

Synonyme: C.I. 45430, Erythrosin 239

C13205000
EHERC13205000EA 46 EUR
EHERC13205000
Erythrosin B Dinatriumsalz
Erythrosin B Dinatriumsalz
Formel: C₂₀H₆I₄Na₂O₅
Molecular Weight: 879,86 g/mol
MDL: MFCD00144257
CAS-Nummer: 16423-68-0
EINECS: 240-474-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR