Ethephon (2-Chlorethylphosphonsäure)

Lieferant: EHRENSTORFER

Synonyme: 2-Chloroethylphosphonic acid, Ethephon, (2-Chloroethyl)phosphonic acid

C13230000
EHERC13230000EA 93 EUR
EHERC13230000
Ethephon (2-Chlorethylphosphonsäure)
Ethephon (2-Chlorethylphosphonsäure)
Formel: C₂H₆ClO₃P
MDL: MFCD00002142
CAS-Nummer: 16672-87-0
EINECS: 240-718-3
UN: 2928
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR