2-Imidazolidinthion
Lieferant: EHRENSTORFER
Synonyme:
2-Mercaptoimidazoline, Ethlenethiourea, Ethylenethiourea, 2-Imidazolidinethione
C13330000
EHERC13330000EA
46
EUR
EHERC13330000
2-Imidazolidinthion
2-Imidazolidinthion
Formel:
C₃H₆N₂S Molecular Weight: 102,16 g/mol |
MDL:
MFCD00005276 CAS-Nummer: 96-45-7 EINECS: 202-506-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...