2-Ethylhexyl p-methoxycinnamate, Dr. Ehrenstorfer
Lieferant: EHRENSTORFER
C13342000
EHERC13342000EA
174
EUR
EHERC13342000
2-Ethylhexyl p-methoxycinnamate, Dr. Ehrenstorfer
Standards
Lebensmittel-Standards
This octinoxate has been used as reference standard for food analysis. It is important in the nutritional and compositional labelling.