2-Chlor-N-(2-ethyl-6-methylphenyl)acetamid

Lieferant: EHRENSTORFER

C13348010
EHERC13348010EA 284 EUR
EHERC13348010
2-Chlor-N-(2-ethyl-6-methylphenyl)acetamid
2-Chlor-N-(2-ethyl-6-methylphenyl)acetamid
Formel: C₁₁H₁₄ClNO
Molecular Weight: 211,6880 g/mol
MDL: MFCD00267498
CAS-Nummer: 32428-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR