Fenclorim
Lieferant: EHRENSTORFER
Synonyme:
4,6-Dichloro-2-phenylpyrimidine
C13463000
EHERC13463000EA
84
EUR
EHERC13463000
Fenclorim
Fenclorim
Formel:
C₁₀H₆Cl₂N₂ Molecular Weight: 225.08 g/mol |
MDL:
MFCD00754193 CAS-Nummer: 3740-92-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...