2-(2,4,5-Trichlorphenoxy)propionsäure
Lieferant: EHRENSTORFER
C13490000
EHERC13490000EA
34
EUR
EHERC13490000
2-(2,4,5-Trichlorphenoxy)propionsäure
2-(2,4,5-Trichlorphenoxy)propionsäure
Formel:
C₉H₇Cl₃O₃ |
MDL:
MFCD00002646 CAS-Nummer: 93-72-1 EINECS: 202-271-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...