Ferbam (Eisentris(dimethyldithiocarbamat))
Lieferant: EHRENSTORFER
Synonyme:
Ferric dimethyldithiocarbamate, Ferbam, Iron(III) dimethyldithiocarbamate, Dimethyldithiocarbamic acid Ferric salt
C13640000
EHERC13640000EA
76
EUR
EHERC13640000
Ferbam (Eisentris(dimethyldithiocarbamat))
Ferbam (Eisentris(dimethyldithiocarbamat))
Formel:
C₉H₁₈FeN₃S₆ |
MDL:
MFCD00067269 CAS-Nummer: 14484-64-1 EINECS: 238-484-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...