Gemfibrozil
Lieferant: EHRENSTORFER
Synonyme:
CI-719, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeriansäure, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentansäure
C13999000
EHERC13999000EA
58
EUR
EHERC13999000
Gemfibrozil
Gemfibrozil
Formel:
C₁₅H₂₂O₃ Molecular Weight: 250,34 g/mol Siedepunkt: 395 °C (1013 hPa) Schmelzpunkt: 59…61 °C |
MDL:
MFCD00079335 CAS-Nummer: 25812-30-0 EINECS: 247-280-2 Merck Index: 12,04394 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...