Guanosin-5'-monophosphorsäure Dinatriumsalz

Lieferant: EHRENSTORFER

Synonyme: Disodium 5'-guanylate, 5'-Guanosine monophosphate disodium salt, GMP disodium salt, 5'-GMP 2Na, 5'-Guanylic acid disodium salt, Guanosine-5'-Monophosphate

C14057150
EHERC14057150EA 122 EUR
EHERC14057150
Guanosin-5'-monophosphorsäure Dinatriumsalz
Guanosin-5'-monophosphorsäure Dinatriumsalz
Formel: C₁₀H₁₂N₅Na₂O₈P
Molecular Weight: 407,187 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00151238
CAS-Nummer: 5550-12-9
EINECS: 226-914-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR