Halofuginone hydrobromide

Lieferant: EHRENSTORFER

Synonyme: 7-Bromo-6-chloro-3-(3-((2S,3R)-rel-3-hydroxypiperidin-2-yl)-2-oxopropyl)quinazolin-4(3H)-one hydrobromide

C14059280
EHERC14059280EA 583 EUR
EHERC14059280
Halofuginone hydrobromide
Halofuginone hydrobromide

Standards: ISO 17034 certified.

Formel: C₁₆H₁₇BrClN₃O₃·HBr
Molecular Weight: 495.59 g/mol
MDL: MFCD00946455
CAS-Nummer: 64924-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR