1-Heneicosanol
Lieferant: EHRENSTORFER
Synonyme:
Heneicosanol, Heneicosyl alcohol, 1-Heneicosanol
C14085200
EHERC14085200EA
78
EUR
EHERC14085200
1-Heneicosanol
1-Heneicosanol
Formel:
H₃C(CH₂)₁₉CH₂OH Molecular Weight: 312.58 g/mol Siedepunkt: 170 Schmelzpunkt: 68…69 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00062834 CAS-Nummer: 15594-90-8 EINECS: 239-673-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...