n-Hentriacontan
Lieferant: EHRENSTORFER
C14085500
EHERC14085500EA
88
EUR
EHERC14085500
n-Hentriacontan
n-Hentriacontane
Formel:
H₃C(CH₂)₂₉CH₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048745 CAS-Nummer: 630-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...