Methylheptadecanoat

Lieferant: EHRENSTORFER

Synonyme: Heptadecansäuremethylester, Methylmargarat, Margarinsäuremethylester

C14122650
EHERC14122650EA 60 EUR
EHERC14122650
Methylheptadecanoat
Methylheptadecanoat
Formel: C₁₈H₃₆O₂
Molecular Weight: 284,48 g/mol
MDL: MFCD00009001
CAS-Nummer: 1731-92-6
EINECS: 217-055-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR