Perchlorethan
Lieferant: EHRENSTORFER
Synonyme:
Hexachloroethane
C14172000
EHERC14172000EA
50
EUR
EHERC14172000
Perchlorethan
Perchlorethan
Formel:
C₂Cl₆ Molecular Weight: 236.74 g/mol Siedepunkt: 189 Dichte: 2.091 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000799 CAS-Nummer: 67-72-1 EINECS: 200-666-4 Merck Index: 13,04698 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...