Hexachlorophen

Lieferant: EHRENSTORFER

Synonyme: 2,2'-Methylenebis(3,4,6-trichlorophenol), 3,3',5,5',6,6'-Hexachloro-2,2'-dihydroxydiphenylmethane

C14180000
EHERC14180000EA 59 EUR
EHERC14180000
Hexachlorophen
Hexachlorophen
Formel: C₁₃H₆Cl₆O₂
Molecular Weight: 406.91 g/mol
MDL: MFCD00002171
CAS-Nummer: 70-30-4
EINECS: 200-733-8
UN: 2875
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR