(±)-2-Hexanol

Lieferant: EHRENSTORFER

Synonyme: (±)-Butyl-methylcarbinol, (±)-Methyl-butylcarbinol, (±)-2-Hydroxyhexan

C14196800
EHERC14196800EA 31 EUR
EHERC14196800
(±)-2-Hexanol
(±)-2-Hexanol
Formel: C₆H₁₄O
Molecular Weight: 102,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004585
CAS-Nummer: 626-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9db7ec887376d519b077785cdead15c1aed20d58&vid=5c0c76fb523ec80ddb5134157d5d19ce7a63dcc3&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR