Hydrocortison-21-acetat

Lieferant: EHRENSTORFER

Synonyme: Hydrocortisone acetate

C14224020
EHERC14224020EA 46 EUR
EHERC14224020
Hydrocortison-21-acetat
Hydrocortison-21-acetat
Formel: C₂₃H₃₂O₆
Molecular Weight: 404,51 g/mol
MDL: MFCD00037714
CAS-Nummer: 50-03-3
EINECS: 200-004-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR