Heptyl-4-hydroxybenzoat

Lieferant: EHRENSTORFER

Synonyme: Heptylparaben, 4-Hydroxybenzoesäureheptylester

C14228820
EHERC14228820EA 70 EUR
EHERC14228820
Heptyl-4-hydroxybenzoat
Heptyl-4-hydroxybenzoat
Formel: C₁₄H₂₀O₃
Molecular Weight: 236,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016481
CAS-Nummer: 1085-12-7
EINECS: 214-115-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR