2-Hydroxy-2-methylpropiophenon

Lieferant: EHRENSTORFER

Synonyme: 2-Benzoyl-2-propanol, 2-Hydroxy-2-methyl-1-phenylpropan-1-one

C14233500
EHERC14233500EA 95 EUR
EHERC14233500
2-Hydroxy-2-methylpropiophenon
2-Hydroxy-2-methylpropiophenon
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2000 g/mol
MDL: MFCD00040710
CAS-Nummer: 7473-98-5
EINECS: 231-272-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR