2-Hydroxy-1,4-naphthochinon

Lieferant: EHRENSTORFER

C14233800
EHERC14233800EA 61 EUR
EHERC14233800
2-Hydroxy-1,4-naphthochinon
2-Hydroxy-1,4-naphthochinon
Formel: C₁₀H₆O₃
MDL: MFCD00001678
CAS-Nummer: 83-72-7
EINECS: 201-496-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR