3-Hydroxy-5-methylisoxazol
Lieferant: EHRENSTORFER
Synonyme:
Hymexazol
C14270000
EHERC14270000EA
126
EUR
EHERC14270000
3-Hydroxy-5-methylisoxazol
3-Hydroxy-5-methylisoxazole
Formel:
C₄H₅NO₂ Molecular Weight: 99,09 g/mol |
MDL:
MFCD00144468 CAS-Nummer: 10004-44-1 EINECS: 233-000-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...