Indigocarmin

Lieferant: EHRENSTORFER
Achtung

Synonyme: Acid Blue 74, Food blue No. 2

C14289000
EHERC14289000EA 51 EUR
EHERC14289000
Indigocarmin
Indigocarmin
Formel: C₁₆H₈N₂Na₂O₈S₂
Molecular Weight: 466,36 g/mol
Schmelzpunkt: > 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005723
CAS-Nummer: 860-22-0
EINECS: 212-728-8
Merck Index: 13,04969

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR