3-Indolylacetonitril
Lieferant: EHRENSTORFER
C14300000
EHERC14300000EA
58
EUR
EHERC14300000
3-Indolylacetonitril
3-Indolylacetonitril
Formel:
C₁₀H₈N₂ |
MDL:
MFCD00005628 CAS-Nummer: 771-51-7 EINECS: 212-232-1 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...