Indometacin
Lieferant: EHRENSTORFER
C14325000
EHERC14325000EA
49
EUR
EHERC14325000
Indometacin
Indomethacin
Formel:
C₁₉H₁₆ClNO₄ Molecular Weight: 357.80 g/mol |
MDL:
MFCD00057095 CAS-Nummer: 53-86-1 EINECS: 200-186-5 UN: 2811 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...