Inosin
Lieferant: EHRENSTORFER
Synonyme:
9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
C14328000
EHERC14328000EA
66
EUR
EHERC14328000
Inosin
Inosin
Formel:
C₁₀H₁₂N₄O₅ Molecular Weight: 268,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066770 CAS-Nummer: 58-63-9 EINECS: 200-390-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...