Isobornylacetat
Lieferant: EHRENSTORFER
C14385000
EHERC14385000EA
44
EUR
EHERC14385000
Isobornylacetat
Isobornylacetat
Formel:
C₁₂H₂₀O₂ |
MDL:
MFCD00135943 CAS-Nummer: 125-12-2 EINECS: 204-727-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...