Methylisobutyrat
Lieferant: EHRENSTORFER
C14396000
EHERC14396000EA
39
EUR
EHERC14396000
Methylisobutyrat
Methylisobutyrat
Formel:
C₅H₁₀O₂ Molecular Weight: 102,13 g/mol |
MDL:
MFCD00008914 CAS-Nummer: 547-63-7 EINECS: 208-929-5 UN: 1237 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...