4-Isopropyl-m-kresol
Lieferant: EHRENSTORFER
Synonyme:
4-Isopropyl-3-methylphenol, 4-Isopropyl-3-hydroxytoluol
C14463650
EHERC14463650EA
86
EUR
EHERC14463650
4-Isopropyl-m-kresol
4-Isopropyl-m-kresol
Formel:
C₁₀H₁₄O Molecular Weight: 150,22 g/mol |
MDL:
MFCD00010704 CAS-Nummer: 3228-02-2 EINECS: 221-761-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...