2-Isopropyl-3-methoxypyrazin

Lieferant: EHRENSTORFER

C14463700
EHERC14463700EA 87 EUR
EHERC14463700
2-Isopropyl-3-methoxypyrazin
2-Isopropyl-3-methoxypyrazin
Formel: C₈H₁₂N₂O
Molecular Weight: 152,1900 g/mol
MDL: MFCD00006135
CAS-Nummer: 25773-40-4
EINECS: 247-256-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR