3-Isopropylphenol
Lieferant: EHRENSTORFER
C14464000
EHERC14464000EA
27.67
EUR
EHERC14464000
3-Isopropylphenol
3-Isopropylphenol
Formel:
C₉H₁₂O Molecular Weight: 136,19 g/mol Siedepunkt: 228 °C (1013 hPa) Schmelzpunkt: 31 °C |
CAS-Nummer:
618-45-1 EINECS: 210-551-0 UN: 2430 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...