5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one, gelb, fest

Lieferant: EHRENSTORFER

Synonyme: Isovitexin

C14479800-10MG
EHERC14479800-10MGEA 227 EUR
EHERC14479800-10MG
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one, gelb, fest
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one
Formel: C₂₁H₂₀O₁₀
Molecular Weight: 432.38 g/mol
Schmelzpunkt: 235...239 °C
MDL: MFCD00076044
CAS-Nummer: 38953-85-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR