Kanamycinsulfat

Lieferant: EHRENSTORFER
Gefahr

Synonyme: Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate

C14505520
EHERC14505520EA 52 EUR
EHERC14505520
Kanamycinsulfat
Kanamycin sulphate
Formel: C₁₈H₃₈N₄O₁₅S
Molecular Weight: 582.6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070253
CAS-Nummer: 25389-94-0
EINECS: 246-933-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR