Ketoprofen

Lieferant: EHRENSTORFER

Synonyme: 2-(3-Benzoylphenyl)propionic acid, 2-(3-Benzoylphenyl)propanoic acid

C14532100
EHERC14532100EA 99 EUR
EHERC14532100
Ketoprofen
Ketoprofen
Formel: C₁₆H₁₄O₃
Molecular Weight: 254,29 g/mol
MDL: MFCD00055790
CAS-Nummer: 22071-15-4
EINECS: 244-759-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR