Malathion (1,2-Bis(ethoxycarbonyl)ethyl-O,O-dimethyldithiophosphat)
Lieferant: EHRENSTORFER
C14710000
EHERC14710000EA
46
EUR
EHERC14710000
Malathion (1,2-Bis(ethoxycarbonyl)ethyl-O,O-dimethyldithiophosphat)
Malathion (1,2-Bis(ethoxycarbonyl)ethyl-O,O-dimethyldithiophosphat)
Formel:
C₁₀H₁₉O₆PS₂ |
CAS-Nummer:
121-75-5 EINECS: 204-497-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...