Melatonin

Lieferant: EHRENSTORFER

Synonyme: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide

C14861500
EHERC14861500EA 58 EUR
EHERC14861500
Melatonin
Melatonin
Formel: C₁₃H₁₆N₂O₂
Molecular Weight: 232,28 g/mol
MDL: MFCD00005655
CAS-Nummer: 73-31-4
EINECS: 200-797-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR