Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3)

Lieferant: EHRENSTORFER

Synonyme: Menadione, 1,4-Dihydro-2-methylnaphthalene-1,4-dione, 2-Methylnaphthalene-1,4-dione

C14863000
EHERC14863000EA 56 EUR
EHERC14863000
Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3)
Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3)
Formel: C₁₁H₈O₂
Molecular Weight: 172,19 g/mol
MDL: MFCD00001681
CAS-Nummer: 58-27-5
EINECS: 200-372-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR