Methomyl (1-Methylthioethylidenamino-N-methylcarbamat)

Lieferant: EHRENSTORFER

C15030000
EHERC15030000EA 51 EUR
EHERC15030000
Methomyl (1-Methylthioethylidenamino-N-methylcarbamat)
Methomyl (1-Methylthioethylidenamino-N-methylcarbamat)
Formel: C₅H₁₀N₂O₂S
CAS-Nummer: 16752-77-5
EINECS: 240-815-0
UN: 2757
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR