4-Methoxy-3,3'-dimethylbenzophenon

Lieferant: EHRENSTORFER

C15081500
EHERC15081500EA 153 EUR
EHERC15081500
4-Methoxy-3,3'-dimethylbenzophenon
4-Methoxy-3,3'-dimethylbenzophenon
Formel: C₁₆H₁₆O₂
MDL: MFCD00128925
CAS-Nummer: 41295-28-7
EINECS: 255-300-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR